Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTGAGCTAACACAGACCTGCTG[A/G]ATCAGCTTCTTTCTGAGCTGCCAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603294 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
C21orf58 - chromosome 21 open reading frame 58 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YBEY - ybeY metallopeptidase (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006114.2 | 764 | UTR 3 | NP_001006114.1 | |||
NM_001314022.1 | 764 | Intron | NP_001300951.1 | |||
NM_001314023.1 | 764 | Intron | NP_001300952.1 | |||
NM_001314024.1 | 764 | Intron | NP_001300953.1 | |||
NM_001314025.1 | 764 | Intron | NP_001300954.1 | |||
NM_001314026.1 | 764 | UTR 3 | NP_001300955.1 | |||
NM_058181.2 | 764 | Intron | NP_478061.1 | |||
XM_005261156.3 | 764 | Intron | XP_005261213.2 | |||
XM_011529629.2 | 764 | Intron | XP_011527931.1 | |||
XM_011529630.2 | 764 | Intron | XP_011527932.1 | |||
XM_011529631.2 | 764 | Intron | XP_011527933.1 | |||
XM_011529633.2 | 764 | Intron | XP_011527935.1 | |||
XM_011529634.2 | 764 | Intron | XP_011527936.2 | |||
XM_011529635.2 | 764 | Intron | XP_011527937.1 | |||
XM_017028394.1 | 764 | Intron | XP_016883883.1 | |||
XM_017028395.1 | 764 | Intron | XP_016883884.1 | |||
XM_017028396.1 | 764 | Intron | XP_016883885.1 |
Set Membership: |
HapMap |