Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACACATCTTTCTCTTCTTCGCC[A/G]CCACCATGGGCACTTTGCTTCAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147150 MIM: 602060 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MX1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MX1 - MX dynamin like GTPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS2 - transmembrane protease, serine 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135099.1 | 2715 | UTR 3 | NP_001128571.1 | |||
NM_005656.3 | 2715 | UTR 3 | NP_005647.3 | |||
XM_005261043.2 | 2715 | UTR 3 | XP_005261100.1 | |||
XM_011529731.2 | 2715 | Intron | XP_011528033.1 | |||
XM_011529732.2 | 2715 | Intron | XP_011528034.1 | |||
XM_011529733.1 | 2715 | UTR 3 | XP_011528035.1 |
Set Membership: |
HapMap |