Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTTGACTTTTGGATAAAGCCTC[C/G]TCCACAGTCCTTGCACACAAAAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF337 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF337 - zinc finger protein 337 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290261.1 | 1270 | Missense Mutation | CGA,GGA | R,G 467 | NP_001277190.1 | |
NM_015655.3 | 1270 | Missense Mutation | CGA,GGA | R,G 467 | NP_056470.1 | |
XM_005260702.3 | 1270 | Nonsense Mutation | CGA,GGA | R,G 435 | XP_005260759.1 | |
XM_006723558.3 | 1270 | Nonsense Mutation | CGA,GGA | R,G 467 | XP_006723621.1 | |
XM_011529219.2 | 1270 | Nonsense Mutation | CGA,GGA | R,G 467 | XP_011527521.1 | |
XM_017027802.1 | 1270 | Nonsense Mutation | CGA,GGA | R,G 435 | XP_016883291.1 | |
XM_017027803.1 | 1270 | Missense Mutation | CGA,GGA | R,G 435 | XP_016883292.1 | |
XM_017027804.1 | 1270 | Nonsense Mutation | CGA,GGA | R,G 402 | XP_016883293.1 |
ZNF337-AS1 - ZNF337 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |