Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAAGACCTTTTTAATATCTAT[C/T]TTTTAGGAGAGATGAATGGAGAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608187 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCM8 - minichromosome maintenance 8 homologous recombination repair factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281520.1 | Intron | NP_001268449.1 | ||||
NM_001281521.1 | Intron | NP_001268450.1 | ||||
NM_001281522.1 | Intron | NP_001268451.1 | ||||
NM_032485.5 | Intron | NP_115874.3 | ||||
NM_182802.2 | Intron | NP_877954.1 | ||||
XM_011529387.2 | Intron | XP_011527689.1 | ||||
XM_017028105.1 | Intron | XP_016883594.1 | ||||
XM_017028106.1 | Intron | XP_016883595.1 | ||||
XM_017028107.1 | Intron | XP_016883596.1 |
TRMT6 - tRNA methyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |