Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTACAACTTTGCAAGCCTGTTT[G/T]CTACTCTTGAACCTAGAATACATTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608187 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCM8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCM8 - minichromosome maintenance 8 homologous recombination repair factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281520.1 | Intron | NP_001268449.1 | ||||
NM_001281521.1 | Intron | NP_001268450.1 | ||||
NM_001281522.1 | Intron | NP_001268451.1 | ||||
NM_032485.5 | Intron | NP_115874.3 | ||||
NM_182802.2 | Intron | NP_877954.1 | ||||
XM_011529387.2 | Intron | XP_011527689.1 | ||||
XM_017028105.1 | Intron | XP_016883594.1 | ||||
XM_017028106.1 | Intron | XP_016883595.1 | ||||
XM_017028107.1 | Intron | XP_016883596.1 |
TRMT6 - tRNA methyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |