Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAAACTTTTTTTAAAAACAGAAA[G/T]AGGAAGGCCTCTCGTACCAGCAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612207 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLPH3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GOLPH3 - golgi phosphoprotein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022130.3 | 1635 | UTR 3 | NP_071413.1 | |||
XM_017009686.1 | 1635 | UTR 3 | XP_016865175.1 | |||
XM_017009687.1 | 1635 | UTR 3 | XP_016865176.1 |