Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTCCTATCTTTGAAGGAAATAA[A/G]TATTTGGTTGCCCTTTTGATGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609803 MIM: 610073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKAR - ankyrin and armadillo repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORMDL1 - ORMDL sphingolipid biosynthesis regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128150.1 | Intron | NP_001121622.1 | ||||
NM_016467.4 | Intron | NP_057551.1 | ||||
XM_005246970.2 | Intron | XP_005247027.1 | ||||
XM_011512199.2 | Intron | XP_011510501.1 | ||||
XM_011512200.2 | Intron | XP_011510502.1 | ||||
XM_017005347.1 | Intron | XP_016860836.1 | ||||
XM_017005348.1 | Intron | XP_016860837.1 |
OSGEPL1 - O-sialoglycoprotein endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSGEPL1-AS1 - OSGEPL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |