Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAAGACATCAGAATGGCTAATA[G/T]TGATTGCATGCTGTGTGTCCACTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605745 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYBRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYBRD1 - cytochrome b reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127383.1 | Intron | NP_001120855.1 | ||||
NM_001256909.1 | Intron | NP_001243838.1 | ||||
NM_024843.3 | Intron | NP_079119.3 |
Set Membership: |
HapMap |