Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATGTTATCGTGGTCAATTCCAG[C/T]CCTTTCCGGCATGTGTTCCACCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608058 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
G6PC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
G6PC2 - glucose-6-phosphatase catalytic subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081686.1 | Intron | NP_001075155.1 | ||||
NM_021176.2 | Intron | NP_066999.1 | ||||
XM_011511564.2 | Intron | XP_011509866.1 | ||||
XM_011511565.2 | Intron | XP_011509867.1 |
Set Membership: |
HapMap |