Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGCTGGAATTAAAGATGACCTG[A/G]TTATGGTTTCCCTTTATTCTCTTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603415 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929680 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929680 - uncharacterized LOC101929680 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN9A - sodium voltage-gated channel alpha subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002977.3 | Intron | NP_002968.1 | ||||
XM_005246757.2 | Intron | XP_005246814.1 | ||||
XM_011511616.2 | Intron | XP_011509918.1 | ||||
XM_011511617.2 | Intron | XP_011509919.1 | ||||
XM_011511618.2 | Intron | XP_011509920.1 | ||||
XM_011511619.2 | Intron | XP_011509921.1 | ||||
XM_017004668.1 | Intron | XP_016860157.1 | ||||
XM_017004669.1 | Intron | XP_016860158.1 |
Set Membership: |
HapMap |