Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAATTAGAGCAAACTGGAAATG[C/T]GCTAATGACGGTTGTATTCTGTAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608136 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF10 - Rho guanine nucleotide exchange factor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308152.1 | Intron | NP_001295081.1 | ||||
NM_001308153.1 | Intron | NP_001295082.1 | ||||
NM_014629.3 | Intron | NP_055444.2 | ||||
XM_005266041.3 | Intron | XP_005266098.1 | ||||
XM_011534767.2 | Intron | XP_011533069.1 | ||||
XM_011534768.1 | Intron | XP_011533070.1 | ||||
XM_011534770.2 | Intron | XP_011533072.1 | ||||
XM_017014003.1 | Intron | XP_016869492.1 |
MIR596 - microRNA 596 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |