Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTCAATACATCCTGTAAGTGTC[A/C]CATAATGGAGTATATTGAACTGAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601114 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MPP3 - membrane palmitoylated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001932.4 | Intron | NP_001923.2 | ||||
XM_006721915.2 | Intron | XP_006721978.1 | ||||
XM_006721916.3 | Intron | XP_006721979.1 | ||||
XM_011524828.2 | Intron | XP_011523130.1 | ||||
XM_017024655.1 | Intron | XP_016880144.1 | ||||
XM_017024656.1 | Intron | XP_016880145.1 | ||||
XM_017024657.1 | Intron | XP_016880146.1 | ||||
XM_017024658.1 | Intron | XP_016880147.1 | ||||
XM_017024659.1 | Intron | XP_016880148.1 | ||||
XM_017024660.1 | Intron | XP_016880149.1 |
Set Membership: |
HapMap |