Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTGTCAAGCTACCTAGCACCAC[A/G]AATAGAACTAATATTAATGGATGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609591 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RIT1 - Ras like without CAAX 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256820.1 | 3043 | UTR 3 | NP_001243749.1 | |||
NM_001256821.1 | 3043 | UTR 3 | NP_001243750.1 | |||
NM_006912.5 | 3043 | UTR 3 | NP_008843.1 |
Set Membership: |
HapMap |