Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTTAGACACATAATTTAGGATC[A/T]TGTCATAAAATGGGTGTAAGTGTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602074 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DAP3 - death associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199849.1 | Intron | NP_001186778.1 | ||||
NM_001199850.1 | Intron | NP_001186779.1 | ||||
NM_001199851.1 | Intron | NP_001186780.1 | ||||
NM_004632.3 | Intron | NP_004623.1 | ||||
NM_033657.2 | Intron | NP_387506.1 | ||||
XM_005245480.2 | Intron | XP_005245537.1 | ||||
XM_005245481.2 | Intron | XP_005245538.1 | ||||
XM_005245482.4 | Intron | XP_005245539.1 | ||||
XM_011509972.2 | Intron | XP_011508274.1 | ||||
XM_017002289.1 | Intron | XP_016857778.1 | ||||
XM_017002290.1 | Intron | XP_016857779.1 | ||||
XM_017002291.1 | Intron | XP_016857780.1 | ||||
XM_017002292.1 | Intron | XP_016857781.1 | ||||
XM_017002293.1 | Intron | XP_016857782.1 | ||||
XM_017002294.1 | Intron | XP_016857783.1 | ||||
XM_017002295.1 | Intron | XP_016857784.1 | ||||
XM_017002296.1 | Intron | XP_016857785.1 | ||||
XM_017002297.1 | Intron | XP_016857786.1 |
Set Membership: |
HapMap |