Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCCATGGAAACTCACCGGGGAA[G/T]AGTCATTTTCTATCGGCTTTGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613890 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD3B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSD3B2 - hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000198.3 | Intron | NP_000189.1 | ||||
NM_001166120.1 | Intron | NP_001159592.1 |
Set Membership: |
HapMap |