Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTAACTTCCTGAATACATTATC[A/G]TCATTATAGTGTAATTAATATTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFP30 - ZFP30 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320666.1 | 3585 | UTR 3 | NP_001307595.1 | |||
NM_001320667.1 | 3585 | UTR 3 | NP_001307596.1 | |||
NM_001320668.1 | 3585 | UTR 3 | NP_001307597.1 | |||
NM_001320669.1 | 3585 | UTR 3 | NP_001307598.1 | |||
NM_001320670.1 | 3585 | UTR 3 | NP_001307599.1 | |||
NM_001320671.1 | 3585 | UTR 3 | NP_001307600.1 | |||
NM_014898.2 | 3585 | UTR 3 | NP_055713.1 |