Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACACCACCTTGTCCAGAGAAAC[A/T]TTTCTAAAGCAGAAGCTGTGGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611053 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL36AP33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RPL36AP33 - ribosomal protein L36a pseudogene 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUSC2 - RUN and SH3 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135999.1 | Intron | NP_001129471.1 | ||||
XM_006716896.2 | Intron | XP_006716959.1 |