Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGACTTTCAAATAATACAGTTTGA[G/T]GTACTCTTGCTTATGTAACCACCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603873 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PLAA - phospholipase A2 activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031689.2 | 3557 | UTR 3 | NP_001026859.1 | |||
NM_001321546.1 | 3557 | UTR 3 | NP_001308475.1 | |||
XM_011518072.2 | 3557 | Intron | XP_011516374.1 | |||
XM_017015319.1 | 3557 | Intron | XP_016870808.1 |
Set Membership: |
HapMap |