Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300403 MIM: 300080 | ||||||||||||||||||||
Literature Links: |
NDUFB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFB11 - NADH:ubiquinone oxidoreductase subunit B11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM10 - RNA binding motif protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204466.1 | Intron | NP_001191395.1 | ||||
NM_001204467.1 | Intron | NP_001191396.1 | ||||
NM_001204468.1 | Intron | NP_001191397.1 | ||||
NM_005676.4 | Intron | NP_005667.2 | ||||
NM_152856.2 | Intron | NP_690595.1 |