Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTCTTACCCGAATGTTGTCATC[A/G]TTGCTGTTGGTTTTCTCCCCTTTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612733 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
THOC5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
THOC5 - THO complex 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002877.1 | 1976 | Silent Mutation | AAC,AAT | N,N 594 | NP_001002877.1 | |
NM_001002878.1 | 1976 | Silent Mutation | AAC,AAT | N,N 594 | NP_001002878.1 | |
NM_001002879.1 | 1976 | Silent Mutation | AAC,AAT | N,N 594 | NP_001002879.1 | |
NM_003678.4 | 1976 | Silent Mutation | AAC,AAT | N,N 594 | NP_003669.4 | |
XM_005261797.1 | 1976 | Silent Mutation | AAC,AAT | N,N 605 | XP_005261854.1 | |
XM_005261798.1 | 1976 | Silent Mutation | AAC,AAT | N,N 605 | XP_005261855.1 | |
XM_005261799.1 | 1976 | Silent Mutation | AAC,AAT | N,N 605 | XP_005261856.1 | |
XM_005261801.3 | 1976 | Silent Mutation | AAC,AAT | N,N 255 | XP_005261858.1 | |
XM_006724347.1 | 1976 | Silent Mutation | AAC,AAT | N,N 605 | XP_006724410.1 | |
XM_017029027.1 | 1976 | Silent Mutation | AAC,AAT | N,N 605 | XP_016884516.1 |