Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGGCTGTGCTGATGGAAAATCG[C/G]CTGTAGTTACATGGGCAAGAATCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612825 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDC4P PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SDC4P - syndecan 4 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC14L4 - SEC14 like lipid binding 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161368.1 | 2717 | Intron | NP_001154840.1 | |||
NM_174977.3 | 2717 | Intron | NP_777637.1 | |||
XM_006724235.2 | 2717 | UTR 3 | XP_006724298.1 | |||
XM_011530164.2 | 2717 | UTR 3 | XP_011528466.1 | |||
XM_017028787.1 | 2717 | UTR 3 | XP_016884276.1 | |||
XM_017028788.1 | 2717 | UTR 3 | XP_016884277.1 | |||
XM_017028789.1 | 2717 | UTR 3 | XP_016884278.1 | |||
XM_017028790.1 | 2717 | Intron | XP_016884279.1 | |||
XM_017028791.1 | 2717 | UTR 3 | XP_016884280.1 |