Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604163 MIM: 611069 MIM: 611068 MIM: 611070 MIM: 611071 | ||||||||||||||||||||
Literature Links: |
RPL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPL3 - ribosomal protein L3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000967.3 | Intron | NP_000958.1 | ||||
NM_001033853.1 | Intron | NP_001029025.1 |
SNORD139 - small nucleolar RNA, C/D box 139 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD43 - small nucleolar RNA, C/D box 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD83A - small nucleolar RNA, C/D box 83A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD83B - small nucleolar RNA, C/D box 83B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |