Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGATGGGACTGGTCCGAGAATC[A/G]GCTTTGACTCCGGGGCTGATTGTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608314 MIM: 610981 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SEPT3 - septin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBP2NL - WBP2 N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152613.2 | Intron | NP_689826.2 |