Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTGATTCCATATACACAGACT[A/G]ATTTTAAGCCATGCATGTAGTCTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611806 MIM: 616600 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AS3MT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AS3MT - arsenite methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BORCS7 - BLOC-1 related complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136200.1 | Intron | NP_001129672.1 | ||||
NM_144591.3 | Intron | NP_653192.2 |
BORCS7-ASMT - BORCS7-ASMT readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |