Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCACTGGTCAGGTCGGGATCCG[C/T]GCCATTAGCATAAAGGCATTTAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD13A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD13A - major facilitator superfamily domain containing 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024789.3 | Intron | NP_079065.2 | ||||
XM_006717973.3 | Intron | XP_006718036.1 | ||||
XM_006717974.3 | Intron | XP_006718037.1 | ||||
XM_011540160.2 | Intron | XP_011538462.1 | ||||
XM_011540163.2 | Intron | XP_011538465.1 | ||||
XM_011540167.2 | Intron | XP_011538469.1 | ||||
XM_011540169.2 | Intron | XP_011538471.1 | ||||
XM_017016654.1 | Intron | XP_016872143.1 | ||||
XM_017016655.1 | Intron | XP_016872144.1 | ||||
XM_017016656.1 | Intron | XP_016872145.1 | ||||
XM_017016657.1 | Intron | XP_016872146.1 | ||||
XM_017016658.1 | Intron | XP_016872147.1 | ||||
XM_017016659.1 | Intron | XP_016872148.1 | ||||
XM_017016660.1 | Intron | XP_016872149.1 | ||||
XM_017016661.1 | Intron | XP_016872150.1 | ||||
XM_017016662.1 | Intron | XP_016872151.1 |
RPARP-AS1 - RPARP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |