Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGTGCAATATTTCAAATATACA[C/G]AAGAGTATCCAGGGGAAAAAAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF145 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF145 - ring finger protein 145 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199380.1 | Intron | NP_001186309.1 | ||||
NM_001199381.1 | Intron | NP_001186310.1 | ||||
NM_001199382.1 | Intron | NP_001186311.1 | ||||
NM_001199383.1 | Intron | NP_001186312.1 | ||||
NM_144726.2 | Intron | NP_653327.1 | ||||
XM_017009138.1 | Intron | XP_016864627.1 |