Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTAATGAGAGATAGGAAAGAGT[C/G]GGGCTGTGACCCCGGTTTAAAGCGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610515 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JADE2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
JADE2 - jade family PHD finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289984.1 | Intron | NP_001276913.1 | ||||
NM_001289985.1 | Intron | NP_001276914.1 | ||||
NM_001308143.1 | Intron | NP_001295072.1 | ||||
NM_015288.5 | Intron | NP_056103.4 | ||||
XM_005271945.1 | Intron | XP_005272002.1 | ||||
XM_005271946.1 | Intron | XP_005272003.1 | ||||
XM_005271948.4 | Intron | XP_005272005.1 | ||||
XM_011543291.2 | Intron | XP_011541593.1 | ||||
XM_017009282.1 | Intron | XP_016864771.1 |
Set Membership: |
HapMap |