Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTTAAAGATATGAATTTTCAT[C/T]TTAGCATAACTTGGTTGCCACAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158378 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC20A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC20A2 - solute carrier family 20 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257180.1 | Intron | NP_001244109.1 | ||||
NM_001257181.1 | Intron | NP_001244110.1 | ||||
NM_006749.4 | Intron | NP_006740.1 | ||||
XM_005273613.3 | Intron | XP_005273670.1 | ||||
XM_005273615.3 | Intron | XP_005273672.1 | ||||
XM_006716390.3 | Intron | XP_006716453.1 | ||||
XM_006716391.3 | Intron | XP_006716454.1 | ||||
XM_017013748.1 | Intron | XP_016869237.1 | ||||
XM_017013749.1 | Intron | XP_016869238.1 | ||||
XM_017013750.1 | Intron | XP_016869239.1 | ||||
XM_017013751.1 | Intron | XP_016869240.1 | ||||
XM_017013752.1 | Intron | XP_016869241.1 |
Set Membership: |
HapMap |