Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGTGGGAGGCGGAGTTAAAGCG[C/T]TAAGAGAAGCGTTACTCCGCGAGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDCA2 - cell division cycle associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317906.1 | 28 | UTR 5 | NP_001304835.1 | |||
NM_001317907.1 | 28 | UTR 5 | NP_001304836.1 | |||
NM_152562.3 | 28 | UTR 5 | NP_689775.2 | |||
XM_006716294.2 | 28 | UTR 5 | XP_006716357.1 | |||
XM_011544415.1 | 28 | UTR 5 | XP_011542717.1 | |||
XM_011544416.2 | 28 | UTR 5 | XP_011542718.1 | |||
XM_011544417.2 | 28 | Intron | XP_011542719.1 | |||
XM_011544418.1 | 28 | UTR 5 | XP_011542720.1 |
KCTD9 - potassium channel tetramerization domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017634.3 | 28 | Intron | NP_060104.2 |