Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTGGAGCGTGGTTTGGAGGCG[C/G]GTGCCCAAGAGTACTGTGAACAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610387 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A37 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC25A37 - solute carrier family 25 member 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317812.1 | Intron | NP_001304741.1 | ||||
NM_001317813.1 | Intron | NP_001304742.1 | ||||
NM_001317814.1 | Intron | NP_001304743.1 | ||||
NM_016612.3 | Intron | NP_057696.2 | ||||
XM_006716352.3 | Intron | XP_006716415.1 | ||||
XM_011544550.2 | Intron | XP_011542852.1 | ||||
XM_011544553.2 | Intron | XP_011542855.1 | ||||
XM_011544554.2 | Intron | XP_011542856.1 | ||||
XM_017013532.1 | Intron | XP_016869021.1 |