Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAGGATTTGTACAAGTTGGCCTC[C/T]TGCCAAATTATCTGGCATTATAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609430 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPAS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPAS3 - neuronal PAS domain protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164749.1 | Intron | NP_001158221.1 | ||||
NM_001165893.1 | Intron | NP_001159365.1 | ||||
NM_022123.2 | Intron | NP_071406.1 | ||||
NM_173159.2 | Intron | NP_775182.1 | ||||
XM_005267991.3 | Intron | XP_005268048.2 | ||||
XM_005267992.3 | Intron | XP_005268049.2 | ||||
XM_011537067.2 | Intron | XP_011535369.1 | ||||
XM_011537069.2 | Intron | XP_011535371.2 | ||||
XM_011537070.2 | Intron | XP_011535372.1 | ||||
XM_011537071.2 | Intron | XP_011535373.2 | ||||
XM_011537072.2 | Intron | XP_011535374.1 | ||||
XM_017021582.1 | Intron | XP_016877071.1 | ||||
XM_017021583.1 | Intron | XP_016877072.1 | ||||
XM_017021584.1 | Intron | XP_016877073.1 | ||||
XM_017021585.1 | Intron | XP_016877074.1 | ||||
XM_017021586.1 | Intron | XP_016877075.1 | ||||
XM_017021587.1 | Intron | XP_016877076.1 | ||||
XM_017021588.1 | Intron | XP_016877077.1 |
Set Membership: |
HapMap |