Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACAAGGCCTCGCGCCGCTGCGG[G/T]TCCTGCGACCGCTCCTGGCTGGTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613317 MIM: 162080 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCAF11 - DDB1 and CUL4 associated factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163484.1 | 127 | UTR 5 | NP_001156956.1 | |||
NM_025230.4 | 127 | UTR 5 | NP_079506.3 | |||
NM_181357.2 | 127 | Intron | NP_852002.1 |
NRL - neural retina leucine zipper | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006177.3 | 127 | Intron | NP_006168.1 | |||
XM_005267708.4 | 127 | UTR 5 | XP_005267765.1 | |||
XM_005267709.3 | 127 | Intron | XP_005267766.1 | |||
XM_005267710.3 | 127 | Intron | XP_005267767.1 | |||
XM_011536801.2 | 127 | Intron | XP_011535103.2 | |||
XM_011536802.1 | 127 | Intron | XP_011535104.1 | |||
XM_011536804.2 | 127 | Intron | XP_011535106.1 | |||
XM_011536805.2 | 127 | Intron | XP_011535107.1 | |||
XM_011536806.2 | 127 | Intron | XP_011535108.2 | |||
XM_017021350.1 | 127 | UTR 5 | XP_016876839.1 |
Set Membership: |
HapMap |