Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATATTTCAGATTTACATATCTTT[C/T]CAGGGATGAATGACTGTCTTAAAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611612 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
THTPA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
THTPA - thiamine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126339.3 | Intron | NP_001119811.1 | ||||
NM_001256062.2 | Intron | NP_001242991.1 | ||||
NM_001256321.2 | Intron | NP_001243250.1 | ||||
NM_001256322.2 | Intron | NP_001243251.1 | ||||
NM_001256323.2 | Intron | NP_001243252.1 | ||||
NM_024328.5 | Intron | NP_077304.1 |
ZFHX2 - zinc finger homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |