Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTATCTCTGTTACTCCTGAAGC[C/G]TCAGAAATTTTAGTCATTATCATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608119 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOMEZ PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HOMEZ - homeobox and leucine zipper encoding | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020834.2 | 1985 | UTR 3 | NP_065885.2 |
RNF212B - ring finger protein 212B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282322.1 | 1985 | UTR 3 | NP_001269251.1 | |||
XM_011536313.2 | 1985 | Intron | XP_011534615.1 | |||
XM_011536314.2 | 1985 | Intron | XP_011534616.1 | |||
XM_011536315.2 | 1985 | Intron | XP_011534617.1 | |||
XM_011536316.2 | 1985 | Intron | XP_011534618.1 | |||
XM_011536319.2 | 1985 | Intron | XP_011534621.1 | |||
XM_011536322.2 | 1985 | Intron | XP_011534624.1 | |||
XM_011536323.2 | 1985 | Intron | XP_011534625.1 | |||
XM_011536324.2 | 1985 | Intron | XP_011534626.1 | |||
XM_011536327.2 | 1985 | Intron | XP_011534629.1 | |||
XM_011536328.2 | 1985 | Intron | XP_011534630.1 | |||
XM_011536334.2 | 1985 | Intron | XP_011534636.1 |
Set Membership: |
HapMap |