Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTACACGGCTATTTGGGAAACT[A/T]AAGTGTTGGAGGAGGCAGGGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615936 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP42 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP42 - Rho GTPase activating protein 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152432.2 | 1004 | Intron | NP_689645.2 | |||
XM_011542615.2 | 1004 | UTR 3 | XP_011540917.1 | |||
XM_011542616.2 | 1004 | UTR 3 | XP_011540918.1 | |||
XM_011542617.2 | 1004 | UTR 3 | XP_011540919.1 | |||
XM_017017237.1 | 1004 | UTR 3 | XP_016872726.1 | |||
XM_017017238.1 | 1004 | UTR 3 | XP_016872727.1 |
TMEM133 - transmembrane protein 133 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032021.2 | 1004 | UTR 3 | NP_114410.1 |
Set Membership: |
HapMap |