Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTCGCTGCCAGGCACAAGTGTCC[C/T]GCCGCATCTCCTTCAGCGCGAGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611740 MIM: 612719 | ||||||||||||||||||||
Literature Links: |
BCO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCO2 - beta-carotene oxygenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTS - 6-pyruvoyltetrahydropterin synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000317.2 | 125 | Missense Mutation | CGC,TGC | R,C 16 | NP_000308.1 |