Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAACCATTATGAACCTTTCCCT[A/G]CATTTCAGTTCCATGGCATATGAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR8G1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OR8G1 - olfactory receptor family 8 subfamily G member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR8G5 - olfactory receptor family 8 subfamily G member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005198.1 | Intron | NP_001005198.1 | ||||
XM_011542662.2 | Intron | XP_011540964.1 | ||||
XM_011542663.2 | Intron | XP_011540965.1 | ||||
XM_011542664.2 | Intron | XP_011540966.1 | ||||
XM_011542665.2 | Intron | XP_011540967.1 | ||||
XM_011542666.2 | Intron | XP_011540968.1 | ||||
XM_011542667.2 | Intron | XP_011540969.1 |
Set Membership: |
HapMap |