Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAAGCAAACAACACAAATACA[G/T]GAAAGCCATTTACTCCTGGTGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603430 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF202 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF202 - zinc finger protein 202 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301779.1 | 3760 | UTR 3 | NP_001288708.1 | |||
NM_001301780.1 | 3760 | UTR 3 | NP_001288709.1 | |||
NM_001301819.1 | 3760 | UTR 3 | NP_001288748.1 | |||
NM_003455.3 | 3760 | UTR 3 | NP_003446.2 | |||
XM_005271659.1 | 3760 | UTR 3 | XP_005271716.1 | |||
XM_005271660.1 | 3760 | UTR 3 | XP_005271717.1 | |||
XM_005271661.1 | 3760 | UTR 3 | XP_005271718.1 | |||
XM_005271664.1 | 3760 | UTR 3 | XP_005271721.1 | |||
XM_006718901.2 | 3760 | UTR 3 | XP_006718964.1 | |||
XM_011542972.1 | 3760 | UTR 3 | XP_011541274.1 | |||
XM_011542973.1 | 3760 | UTR 3 | XP_011541275.1 | |||
XM_011542975.1 | 3760 | UTR 3 | XP_011541277.1 | |||
XM_011542976.1 | 3760 | UTR 3 | XP_011541278.1 | |||
XM_017018268.1 | 3760 | UTR 3 | XP_016873757.1 |
Set Membership: |
HapMap |