Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAGCTGGGGCCAAGAGCCTGTT[G/T]TATGAGCGGGTGGACCTGGGAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDGFRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HDGFRP2 - hepatoma-derived growth factor-related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001520.2 | Intron | NP_001001520.1 | ||||
NM_032631.3 | Intron | NP_116020.1 | ||||
XM_011528375.2 | Intron | XP_011526677.1 | ||||
XM_011528376.2 | Intron | XP_011526678.1 | ||||
XM_011528377.2 | Intron | XP_011526679.1 |
Set Membership: |
HapMap |