Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGGCGGCGGCGGCGGGGCCGG[A/G]GCGGGGGGCTGCGGCGGGCCCGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607881 | ||||||||||||||||||||
Literature Links: |
MPND PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPND - MPN domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159846.2 | 244 | Silent Mutation | GGA,GGG | G,G 59 | NP_001153318.1 | |
NM_001300862.1 | 244 | Silent Mutation | GGA,GGG | G,G 59 | NP_001287791.1 | |
NM_032868.5 | 244 | Silent Mutation | GGA,GGG | G,G 59 | NP_116257.2 | |
XM_006722926.2 | 244 | Silent Mutation | GGA,GGG | G,G 59 | XP_006722989.1 |
STAP2 - signal transducing adaptor family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |