Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGTATGGGGGAGGGTGCAGAA[A/G]AAAACCTGGCTCCTCTTCTTGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606597 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC140 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC140 - coiled-coil domain containing 140 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153038.1 | Intron | NP_694583.1 | ||||
XM_017003466.1 | Intron | XP_016858955.1 |
PAX3 - paired box 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000438.5 | Intron | NP_000429.2 | ||||
NM_001127366.2 | Intron | NP_001120838.1 | ||||
NM_013942.4 | Intron | NP_039230.1 | ||||
NM_181457.3 | Intron | NP_852122.1 | ||||
NM_181458.3 | Intron | NP_852123.1 | ||||
NM_181459.3 | Intron | NP_852124.1 | ||||
NM_181460.3 | Intron | NP_852125.1 | ||||
NM_181461.3 | Intron | NP_852126.1 |
Set Membership: |
HapMap |