Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCTGGTTCTTCCTGTGAACAC[A/C]CCCCATTTAGTATTTCAGCTACCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 177010 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SERPINE2 - serpin family E member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136528.1 | Intron | NP_001130000.1 | ||||
NM_001136530.1 | Intron | NP_001130002.1 | ||||
NM_006216.3 | Intron | NP_006207.1 | ||||
XM_005246641.2 | Intron | XP_005246698.1 | ||||
XM_017004329.1 | Intron | XP_016859818.1 | ||||
XM_017004330.1 | Intron | XP_016859819.1 | ||||
XM_017004331.1 | Intron | XP_016859820.1 | ||||
XM_017004332.1 | Intron | XP_016859821.1 |
Set Membership: |
HapMap |