Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCTTTCTGTCATACCTTATGA[C/T]GTGCTGGTCTCTCCCAGGGTTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616010 MIM: 602676 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPS7B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COPS7B - COP9 signalosome subunit 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282949.2 | Intron | NP_001269878.1 | ||||
NM_001282950.2 | Intron | NP_001269879.1 | ||||
NM_001282951.2 | Intron | NP_001269880.1 | ||||
NM_001282952.2 | Intron | NP_001269881.1 | ||||
NM_001308381.1 | Intron | NP_001295310.1 | ||||
NM_022730.3 | Intron | NP_073567.1 | ||||
XM_006712693.1 | Intron | XP_006712756.1 | ||||
XM_011511638.1 | Intron | XP_011509940.1 | ||||
XM_011511639.1 | Intron | XP_011509941.1 | ||||
XM_011511640.1 | Intron | XP_011509942.1 | ||||
XM_017004716.1 | Intron | XP_016860205.1 | ||||
XM_017004717.1 | Intron | XP_016860206.1 |
PDE6D - phosphodiesterase 6D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |