Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGAAACCACTGAAGTGATTCATC[A/G]TACCTGAACGTTGCTGGATGGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5DC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NT5DC4 - 5'-nucleotidase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011512262.2 | 1263 | Intron | XP_011510564.1 | |||
XM_011512265.2 | 1263 | Intron | XP_011510567.1 | |||
XM_017005474.1 | 1263 | Intron | XP_016860963.1 | |||
XM_017005475.1 | 1263 | Intron | XP_016860964.1 | |||
XM_017005476.1 | 1263 | Intron | XP_016860965.1 | |||
XM_017005477.1 | 1263 | UTR 5 | XP_016860966.1 | |||
XM_017005478.1 | 1263 | Intron | XP_016860967.1 |
Set Membership: |
HapMap |