Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTACATAAAATTTAAATGGGTTG[A/C]TAGATCCCCTGAAGTGTATCCACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147679 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL1RN - interleukin 1 receptor antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000577.4 | Intron | NP_000568.1 | ||||
NM_001318914.1 | Intron | NP_001305843.1 | ||||
NM_173841.2 | Intron | NP_776213.1 | ||||
NM_173842.2 | Intron | NP_776214.1 | ||||
NM_173843.2 | Intron | NP_776215.1 | ||||
XM_005263661.4 | Intron | XP_005263718.1 | ||||
XM_011511121.1 | Intron | XP_011509423.1 |
Set Membership: |
HapMap |