Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCCGCCCTCAGGCTATATAAG[C/T]GCACTCGCGGCGGCGTTGGTGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614030 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPDYC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPDYC - speedy/RINGO cell cycle regulator family member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008778.1 | Intron | NP_001008778.1 | ||||
XM_017017705.1 | Intron | XP_016873194.1 | ||||
XM_017017706.1 | Intron | XP_016873195.1 | ||||
XM_017017707.1 | Intron | XP_016873196.1 | ||||
XM_017017708.1 | Intron | XP_016873197.1 | ||||
XM_017017709.1 | Intron | XP_016873198.1 | ||||
XM_017017710.1 | Intron | XP_016873199.1 |