Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134690 MIM: 606866 MIM: 603414 MIM: 615738 MIM: 604575 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAU PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAU - FAU, ubiquitin like and ribosomal protein S30 fusion | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL49 - mitochondrial ribosomal protein L49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM7SF2 - transmembrane 7 superfamily member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277233.1 | Intron | NP_001264162.1 | ||||
NM_003273.3 | Intron | NP_003264.2 |
VPS51 - VPS51, GARP complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNHIT2 - zinc finger HIT-type containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |