Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCCCACCTCACCCGTACCCTC[A/G]CTACCTCATGCCGGAGGCTGCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136425 MIM: 600829 MIM: 602753 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOLR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
FOLR2 - folate receptor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INPPL1 - inositol polyphosphate phosphatase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001567.3 | 1005 | Missense Mutation | ACT,GCT | T,A 212 | NP_001558.3 | |
XM_005273978.3 | 1005 | Missense Mutation | ACT,GCT | T,A 234 | XP_005274035.1 | |
XM_005273979.4 | 1005 | Missense Mutation | ACT,GCT | T,A 234 | XP_005274036.1 | |
XM_011544999.2 | 1005 | Missense Mutation | ACT,GCT | T,A 212 | XP_011543301.1 | |
XM_011545000.2 | 1005 | Missense Mutation | ACT,GCT | T,A 234 | XP_011543302.1 | |
XM_017017673.1 | 1005 | Missense Mutation | ACT,GCT | T,A 224 | XP_016873162.1 |
PHOX2A - paired like homeobox 2a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |