Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGATGAGGGCAGAGATGCCAAAC[G/T]GCCTGCGCCCATTGCTCTGCGTATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606607 MIM: 606472 | ||||||||||||||||||||
Literature Links: |
LSM14B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LSM14B - LSM family member 14B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMA7 - proteasome subunit alpha 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002792.3 | 532 | Missense Mutation | CAG,CCG | Q,P 126 | NP_002783.1 |
SS18L1 - SS18L1, nBAF chromatin remodeling complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |