Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTGGGTTCAAAATTTCAACGA[A/T]ACTGAATGAGTCCCGCGGCGGGTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605917 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TPX2 - TPX2, microtubule nucleation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012112.4 | 39 | UTR 5 | NP_036244.2 | |||
XM_011528697.2 | 39 | UTR 5 | XP_011526999.1 | |||
XM_011528699.2 | 39 | UTR 5 | XP_011527001.1 | |||
XM_011528700.2 | 39 | Intron | XP_011527002.1 |
Set Membership: |
HapMap |